Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200227.1 | NP_956521.1 | 1 - 2 | 162 | 80 | |
GenBank mRNA | BC046064.1 | - | 1 - 2 | 162 | 80 | |
BC071377.1 | - | 1 - 2 | 198 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |