Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213319.2 | NP_998484.1 | 8 - 9 | 1407 | 76 | |
GenBank mRNA | BC057409.1 | - | 8 - 9 | 1407 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |