Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044913.1 | NP_001038378.1 | 15 - 16 | 2158 | 70 | |
GenBank mRNA | BC133137.1 | - | 15 - 16 | 2232 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |