Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131418.2 | NP_571493.2 | 12 - 13 | 1638 | 77 | |
XM_005165618.4 | XP_005165675.1 | 12 - 13 | 1690 | 77 | ||
GenBank mRNA | AF072657.1 | - | 12 - 13 | 77 | ||
BC162504.1 | - | 12 - 13 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |