Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199659.2 | NP_955953.2 | 2 - 3 | 473 | 78 | |
XM_021478841.1 | XP_021334516.1 | 2 - 3 | 501 | 78 | ||
GenBank mRNA | BC045445.1 | - | 2 - 3 | 457 | 78 | |
BC066480.1 | - | 2 - 3 | 473 | 78 | ||
BC165664.1 | - | 1 - 2 | 172 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |