Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptk2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212570.1 | NP_997735.1 | 24 - 25 | 2557 | 86 | |
GenBank mRNA | AJ490510.2 | - | 24 - 25 | 2557 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |