Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anp32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212603.2 | NP_997768.2 | 1 - 2 | 213 | 82 | |
XM_005159865.4 | XP_005159922.1 | 1 - 2 | 1441 | 82 | ||
GenBank mRNA | BC065856.1 | - | 1 - 2 | 158 | 82 | |
CV118158.1 | - | 1 - 2 | 164 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |