Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dytn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080587.1 | NP_001074056.1 | 7 - 8 | 755 | 64 | |
XM_009302235.3 | XP_009300510.1 | 8 - 9 | 1036 | 64 | ||
XM_009302236.3 | XP_009300511.1 | 8 - 9 | 1033 | 64 | ||
GenBank mRNA | DQ443726.1 | - | 7 - 8 | 755 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |