Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131033.2 | NP_571108.2 | 5 - 6 | 717 | 74 | |
GenBank mRNA | AJ249796.1 | - | 5 - 6 | 417 | 74 | |
BC122249.1 | - | 5 - 6 | 717 | 74 | ||
EH513303.1 | - | 5 - 6 | 476 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |