Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fth1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131585.1 | NP_571660.1 | 2 - 3 | 317 | 86 | |
XM_017356903.2 | XP_017212392.1 | 3 - 4 | 444 | 86 | ||
GenBank mRNA | AF295373.1 | - | 2 - 3 | 317 | 86 | |
BC045278.1 | - | 2 - 3 | 350 | 86 | ||
BC163940.1 | - | 1 - 2 | 137 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |