Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131714.2 | NP_571789.2 | 8 - 9 | 1109 | 132 | |
XM_005173133.2 | XP_005173190.1 | 6 - 7 | 1066 | 132 | ||
GenBank mRNA | AB032415.1 | - | 8 - 9 | 1162 | 132 | |
BC045280.1 | - | 8 - 9 | 1109 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |