Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201063.1 | NP_957357.1 | 1 - 2 | 860 | 80 | |
GenBank mRNA | BC054898.1 | - | 1 - 2 | 860 | 80 | |
CU468779.1 | - | 1 - 2 | 787 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |