Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY077715.1 | - | 3 - 4 | 382 | 62 | |
BC050484.1 | - | 3 - 4 | 383 | 62 | ||
BC067161.1 | - | 3 - 4 | 332 | 62 | ||
BC152191.1 | - | 3 - 4 | 396 | 62 | ||
CF996108.1 | - | 3 - 4 | 337 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |