Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213349.1 | NP_998514.1 | 4 - 5 | 417 | 106 | |
GenBank mRNA | BC059802.1 | - | 4 - 5 | 417 | 106 | |
DQ104245.1 | - | 4 - 5 | 423 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |