Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: creg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007305.1 | NP_001007306.1 | 3 - 4 | 734 | 69 | |
GenBank mRNA | BC085654.1 | - | 3 - 4 | 734 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |