Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017773.1 | NP_001017773.1 | 7 - 8 | 835 | 73 | |
GenBank mRNA | BC092907.1 | - | 7 - 8 | 835 | 73 | |
BC152126.1 | - | 7 - 8 | 744 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |