Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dvl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044917.1 | NP_001038382.1 | 5 - 6 | 586 | 112 | |
XM_005169297.4 | XP_005169354.1 | 5 - 6 | 1004 | 112 | ||
XM_005169298.4 | XP_005169355.1 | 5 - 6 | 1004 | 112 | ||
XM_005169299.4 | XP_005169356.1 | 3 - 4 | 822 | 112 | ||
XM_005169301.4 | XP_005169358.1 | 5 - 6 | 1004 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |