Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045214.1 | NP_001038679.1 | 16 - 17 | 1807 | 94 | |
XM_005170062.4 | XP_005170119.1 | 16 - 17 | 2218 | 94 | ||
GenBank mRNA | BC163250.1 | - | 15 - 16 | 1765 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |