Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313682.1 | NP_001300611.1 | 5 - 6 | 921 | 87 | |
XM_021475995.1 | XP_021331670.1 | 5 - 6 | 620 | 87 | ||
GenBank mRNA | AF152607.1 | - | 5 - 6 | 674 | 87 | |
AF179407.1 | - | 5 - 6 | 917 | 87 | ||
AF185283.1 | - | 4 - 5 | 419 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |