Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tenm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130969.2 | NP_571044.2 | 12 - 13 | 2513 | 89 | |
XM_005157483.4 | XP_005157540.1 | 12 - 13 | 2585 | 89 | ||
XM_009291504.3 | XP_009289779.1 | 12 - 13 | 2585 | 89 | ||
XM_009291505.3 | XP_009289780.1 | 12 - 13 | 2585 | 89 | ||
XM_009291506.3 | XP_009289781.1 | 12 - 13 | 2561 | 89 | ||
XM_009291508.3 | XP_009289783.1 | 11 - 12 | 2486 | 89 | ||
XM_009291509.3 | XP_009289784.1 | 11 - 12 | 2486 | 89 | ||
XM_009291510.3 | XP_009289785.1 | 11 - 12 | 2365 | 89 | ||
XM_009291511.3 | XP_009289786.1 | 11 - 12 | 2485 | 89 | ||
GenBank mRNA | AB026980.1 | - | 12 - 13 | 2512 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |