Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199748.1 | NP_956042.1 | 4 - 5 | 584 | 86 | |
XM_005158720.4 | XP_005158777.1 | 4 - 5 | 793 | 86 | ||
GenBank mRNA | BC053169.1 | - | 4 - 5 | 584 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |