Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk1db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213250.1 | NP_998415.1 | 9 - 10 | 1637 | 85 | |
GenBank mRNA | BC063953.1 | - | 9 - 10 | 1637 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |