Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxylp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002430.1 | NP_001002430.1 | 2 - 3 | 260 | 58 | |
XM_005157794.4 | XP_005157851.1 | 2 - 3 | 254 | 58 | ||
GenBank mRNA | BC076136.1 | - | 2 - 3 | 260 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |