Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017801.1 | NP_001017801.1 | 4 - 5 | 570 | 69 | |
XM_005159741.1 | XP_005159798.1 | 4 - 5 | 617 | 69 | ||
GenBank mRNA | BC092880.1 | - | 4 - 5 | 570 | 69 | |
BC164311.1 | - | 3 - 4 | 463 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |