Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045381.2 | NP_001038846.2 | 4 - 5 | 914 | 102 | |
GenBank mRNA | BC122370.1 | - | 4 - 5 | 901 | 102 | |
EH572300.1 | - | 1 - 2 | 238 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |