Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130973.3 | NP_571048.2 | 18 - 19 | 2815 | 132 | |
GenBank mRNA | AF034606.1 | - | 18 - 19 | 2817 | 132 | |
BC162594.1 | - | 18 - 19 | 2817 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |