Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: her7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131609.1 | NP_571684.1 | 1 - 2 | 145 | 91 | |
GenBank mRNA | AF240772.1 | - | 1 - 2 | 145 | 91 | |
BC092709.1 | - | 1 - 2 | 168 | 91 | ||
BC163871.1 | - | 1 - 2 | 144 | 91 | ||
BC163885.1 | - | 1 - 2 | 144 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |