Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200238.1 | NP_956532.1 | 6 - 7 | 647 | 70 | |
XM_005167920.4 | XP_005167977.1 | 6 - 7 | 725 | 70 | ||
GenBank mRNA | BC047186.1 | - | 6 - 7 | 647 | 70 | |
BC165692.1 | - | 5 - 6 | 600 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |