Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf2h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128422.1 | NP_001121894.1 | 9 - 10 | 717 | 125 | |
GenBank mRNA | BC076236.1 | - | 9 - 10 | 689 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |