Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbpja | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198878.2 | NP_942579.2 | 2 - 3 | 83 | 70 | |
GenBank mRNA | AY128532.1 | - | 2 - 3 | 83 | 70 | |
AY332617.1 | - | 2 - 3 | 56 | 70 | ||
BC092918.1 | - | 2 - 3 | 60 | 70 | ||
BC163922.1 | - | 2 - 3 | 83 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |