Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim55a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002358.1 | NP_001002358.1 | 7 - 8 | 1000 | 66 | |
GenBank mRNA | BC075897.1 | - | 7 - 8 | 1000 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |