Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004550.1 | NP_001004550.1 | 2 - 3 | 460 | 62 | |
XM_005173951.4 | XP_005174008.1 | 3 - 4 | 1102 | 62 | ||
XM_017353113.2 | XP_017208602.1 | 3 - 4 | 821 | 62 | ||
GenBank mRNA | BC081661.1 | - | 2 - 3 | 460 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |