Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 12 - 13 | 1453 | 56 | |
XM_005160684.4 | XP_005160741.1 | 11 - 12 | 1084 | 56 | ||
XM_009294709.3 | XP_009292984.1 | 12 - 13 | 1484 | 56 | ||
GenBank mRNA | DT075242.1 | - | 1 - 2 | 50 | 56 | |
DT081164.1 | - | 7 - 8 | 573 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |