Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 9 - 10 | 1300 | 91 | |
XM_005160684.4 | XP_005160741.1 | 8 - 9 | 931 | 91 | ||
XM_009294709.3 | XP_009292984.1 | 9 - 10 | 1331 | 91 | ||
GenBank mRNA | DT081164.1 | - | 4 - 5 | 420 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |