Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnx2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009887.2 | NP_001009887.2 | 1 - 2 | 500 | 63 | |
GenBank mRNA | AY445046.1 | - | 1 - 2 | 497 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |