Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017831.1 | NP_001017831.1 | 4 - 5 | 584 | 77 | |
XM_005173119.4 | XP_005173176.1 | 4 - 5 | 621 | 77 | ||
GenBank mRNA | BC092784.1 | - | 4 - 5 | 584 | 77 | |
BC165264.1 | - | 3 - 4 | 475 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |