Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077449.1 | NP_001070917.1 | 38 - 39 | 6561 | 83 | |
XM_005167736.4 | XP_005167793.1 | 39 - 40 | 7061 | 83 | ||
XM_009304755.3 | XP_009303030.1 | 39 - 40 | 7062 | 83 | ||
XM_009304756.3 | XP_009303031.1 | 39 - 40 | 7062 | 83 | ||
XM_009304757.3 | XP_009303032.1 | 39 - 40 | 7063 | 83 | ||
GenBank mRNA | DQ086492.1 | - | 38 - 39 | 6561 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |