Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080193.1 | NP_001073662.1 | 11 - 12 | 1226 | 89 | |
XM_005168796.4 | XP_005168853.1 | 11 - 12 | 3927 | 89 | ||
GenBank mRNA | BC066564.1 | - | 11 - 12 | 1545 | 89 | |
BC129198.1 | - | 11 - 12 | 1226 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |