Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hcfc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045064.1 | NP_001038529.1 | 21 - 22 | 4402 | 91 | |
XM_005169304.4 | XP_005169361.1 | 21 - 22 | 4489 | 91 | ||
XM_009306034.3 | XP_009304309.1 | 21 - 22 | 4469 | 91 | ||
XM_009306035.3 | XP_009304310.1 | 21 - 22 | 4462 | 91 | ||
GenBank mRNA | BC139848.1 | - | 21 - 22 | 4407 | 91 | |
BC142544.1 | - | 21 - 22 | 4407 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |