Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045074.1 | NP_001038539.1 | 3 - 4 | 618 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |