Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089476.1 | NP_001082945.1 | 2 - 3 | 523 | 92 | |
GenBank mRNA | BC139552.1 | - | 2 - 3 | 523 | 92 | |
BC171418.1 | - | 1 - 2 | 513 | 92 | ||
BC171424.1 | - | 1 - 2 | 513 | 92 | ||
FJ969446.1 | - | 2 - 3 | 556 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |