Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rock2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174863.2 | NP_777288.1 | 18 - 19 | 2606 | 130 | |
GenBank mRNA | AF295804.1 | - | 18 - 19 | 2606 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |