Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zmynd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183069.1 | NP_898892.1 | 5 - 6 | 687 | 65 | |
GenBank mRNA | BC045296.1 | - | 5 - 6 | 687 | 65 | |
BC165425.1 | - | 5 - 6 | 576 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |