Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rasgef1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199829.1 | NP_956123.1 | 9 - 10 | 1235 | 59 | |
XM_005165278.4 | XP_005165335.1 | 10 - 11 | 1362 | 59 | ||
XM_005165279.4 | XP_005165336.1 | 9 - 10 | 1311 | 59 | ||
XM_005165280.4 | XP_005165337.1 | 8 - 9 | 1209 | 59 | ||
GenBank mRNA | BC055678.1 | - | 9 - 10 | 1235 | 59 | |
BC075904.1 | - | 8 - 9 | 1175 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |