Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc10a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200358.1 | NP_956652.1 | 1 - 2 | 548 | 66 | |
GenBank mRNA | BC053189.1 | - | 1 - 2 | 548 | 66 | |
BC165330.1 | - | 1 - 2 | 415 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |