Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jade3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201652.2 | NP_970614.2 | 10 - 11 | 1646 | 93 | |
GenBank mRNA | BC045468.1 | - | 10 - 11 | 1646 | 93 | |
BC165248.1 | - | 8 - 9 | 1467 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |