Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014816.1 | NP_001014816.1 | 3 - 4 | 912 | 71 | |
XM_005166379.4 | XP_005166436.1 | 4 - 5 | 1083 | 71 | ||
GenBank mRNA | AY954727.1 | - | 3 - 4 | 912 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons |