Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040687.1 | NP_001035777.1 | 26 - 27 | 3403 | 73 | |
XM_017351014.2 | XP_017206503.1 | 28 - 29 | 3995 | 73 | ||
XM_021481465.1 | XP_021337140.1 | 27 - 28 | 3879 | 73 | ||
GenBank mRNA | DQ466904.1 | - | 26 - 27 | 3403 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |