Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131676.2 | NP_571751.1 | 3 - 4 | 318 | 81 | |
GenBank mRNA | AF155576.1 | - | 3 - 4 | 320 | 81 | |
BC076475.1 | - | 3 - 4 | 333 | 81 | ||
BC165038.1 | - | 3 - 4 | 280 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |