Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199653.1 | NP_955947.1 | 7 - 8 | 1078 | 63 | |
GenBank mRNA | BC053153.1 | - | 7 - 8 | 1078 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |